Invitrogen
Manufacturer Code:PA542782
Catalog # PIPA542782
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human N6AMT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HemK methyltransferase family member 2; Lysine N-methyltransferase 9; M-N6DNA MTase A; M.HsaHemK2P; Methylarsonite methyltransferase N6AMT1; Methyltransferase N6AMT1; N(6)-adenine-specific DNA methyltransferase 1; N(6)-adenine-specific DNA methyltransferase 1-like; N-6 adenine-specific DNA methyltransferase 1 (putative); N6-DNA methyltransferase A; N6-DNA-methyltransferase; Protein N(5)-glutamine methyltransferase
Gene Aliases: 5830445C04Rik; C21orf127; HEMK2; KMT9; m.HsaHemK2P; MTQ2; N6AMT; N6AMT1; PRED28; RGD1311843
UniProt ID: (Human) Q9Y5N5, (Mouse) Q6SKR2
Entrez Gene ID: (Horse) 100065688, (Human) 29104, (Bovine) 615200, (Rabbit) 100352597, (Guinea pig) 100714564, (Mouse) 67768, (Rat) 288309
Molecular Function: DNA binding protein DNA methyltransferase RNA binding protein RNA methyltransferase methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.