Abnova
Manufacturer Code:H00029104K
Catalog # 89-202-076
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human N6AMT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human N6AMT1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: C21orf127 HEMK2 MGC19995 MTQ2 N6AMT PRED28; HemK methyltransferase family member 2; Lysine N-methyltransferase 9; M.HsaHemK2P; Methylarsonite methyltransferase N6AMT1; Methyltransferase N6AMT1; n(6)-adenine-specific DNA methyltransferase 1; N6-DNA-methyltransferase; Protein N(5)-glutamine methyltransferase
Gene Aliases: C21orf127; HEMK2; KMT9; m.HsaHemK2P; MTQ2; N6AMT; N6AMT1; PRED28
UniProt ID: (Human) Q9Y5N5
Entrez Gene ID: (Human) 29104
Molecular Function:
DNA binding protein
DNA methyltransferase
RNA binding protein
RNA methyltransferase
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.