Proteintech
Manufacturer Code:167331AP150UL
Catalog # 50-173-1147
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | N4BP3 Fusion Protein Ag10194 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0341 N4BP3 Nedd4 binding protein 3; N4BP3; N4BP3 {ECO:0000250|UniProtKB:O15049}; N4bp3 {ECO:0000312|RGD:1308356}; Nedd4 binding protein 3; nedd4 binding protein 3 {ECO:0000312|EMBL:ABA06436.1}; NEDD4-binding protein 3
Gene Aliases: C330016O10Rik; KIAA0341; LZTS4; N4BP3; RGD1308356
UniProt ID: (Human) O15049, (Rat) Q3LUD3, (Mouse) Q8C7U1
Entrez Gene ID: (Human) 23138, (Rat) 303112, (Mouse) 212706
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.