Invitrogen
Manufacturer Code:PA539341
Catalog # PIPA539341
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human N4BP2L2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NEDD4 binding protein 2-like 2; NEDD4-binding protein 2-like 2; NEDD4-binding protein 2-like 2 Phosphonoformate immuno-associated protein 5 CG005 PFAAP5; Phosphonoformate immuno-associated protein 5; Phosphonoformate immuno-associated protein 5 homolog; protein from BRCA2 region
Gene Aliases: 2700092H06Rik; 92M18.3; CG005; CG016; N4BP2L2; PFAAP5; zag1
UniProt ID: (Human) Q92802, (Mouse) Q8JZS6
Entrez Gene ID: (Human) 10443, (Mouse) 381695
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.