Invitrogen
Manufacturer Code:PA5110009
Catalog # PIPA5110009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 40-120 of human N4BP2L1 (NP_001073159). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410024N18Rik AI428195 B230342M21Rik CG018 CG081 N4bp2l1 NEDD4-binding protein 2-like 1; hypothetical gene CG018; NEDD4 binding protein 2-like 1; NEDD4-binding protein 2-like 1
Gene Aliases: 2410024N18Rik; AI428195; B230342M21Rik; CG018; CG081; N4BP2L1
UniProt ID: (Human) Q5TBK1, (Mouse) Q3V2Q8
Entrez Gene ID: (Human) 90634, (Mouse) 100637
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.