Novus Biologicals
Manufacturer Code:NBP2832500.1ML
Catalog # NB031179
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human N4BP2L1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CG018 CG081 NEDD4 binding protein 2-like 1 NEDD4-binding protein 2-like 1; hypothetical gene CG018; NEDD4 binding protein 2-like 1; NEDD4-binding protein 2-like 1
Gene Aliases: CG018; CG081; N4BP2L1
UniProt ID: (Human) Q5TBK1
Entrez Gene ID: (Human) 90634
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.