Novus Biologicals
Manufacturer Code:NBP298059UV
Catalog # NB051889
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human N4BP2 fragment. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B3BPKIAA1413 BCL-3-binding protein EC 3.- FLJ10680 NEDD4 binding protein 2 NEDD4-binding protein 2; BCL-3-binding protein; N4BP2; NEDD4-binding protein 2
Gene Aliases: B3BP; KIAA1413; N4BP2
UniProt ID: (Human) Q86UW6
Entrez Gene ID: (Human) 55728
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.