Novus Biologicals
Manufacturer Code:NBP237688
Catalog # NBP237688
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody maps to a region between residue 250 to 300 of human NEDD4 Binding Protein 1 using the numbering given in entry NP_694574.3 (GeneID 9683). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ31821 KIAA0615 MGC176730 NEDD4 binding protein 1 NEDD4-binding protein 1; N4BP1; NEDD4-binding protein 1
Gene Aliases: KIAA0615; N4BP1
UniProt ID: (Human) O75113
Entrez Gene ID: (Human) 9683
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.