Invitrogen
Manufacturer Code:MA524328
Catalog # PIMA524328
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 709116 |
Immunogen | Mouse myeloma cell line NS0-derived recombit human ULBP-4/RAET1E Gly30-Asp225 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: LETAL N2DL4 N2DL-4 NKG2DL4 RAET1E RL-4 bA350J20.7 LETALRAE-1-like transcript 4 Lymphocyte effector toxicity activation ligand MGC125309 N2DL4 N2DL-4 NKG2D ligand 4 NKG2DL4 RAET1E2 retinoic acid early transcript 1ERL-4 ULBP4MGC125308; Lymphocyte effector toxicity activation ligand; N2DL-4; NKG2D ligand 4; RAE-1-like transcript 4; Retinoic acid early transcript 1E; UL16-binding protein 4
Gene Aliases: bA350J20.7; LETAL; N2DL-4; N2DL4; NKG2DL4; RAET1E; RAET1E2; RL-4; ULBP4; UNQ1867/PRO4303
UniProt ID: (Human) Q8TD07
Entrez Gene ID: (Human) 135250
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
major histocompatibility complex antigen
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.