Invitrogen
Manufacturer Code:PA584180
Catalog # PIPA584180
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RAET1E |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA350J20.7 LETAL ULBP4; Lymphocyte effector toxicity activation ligand; N2DL-4; NKG2D ligand 4; RAE-1-like transcript 4; Retinoic acid early transcript 1E; UL16-binding protein 4
Gene Aliases: bA350J20.7; LETAL; N2DL-4; N2DL4; NKG2DL4; RAET1E; RAET1E2; RL-4; ULBP4; UNQ1867/PRO4303
UniProt ID: (Human) Q8TD07
Entrez Gene ID: (Human) 135250
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily major histocompatibility complex antigen receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.