Invitrogen
Manufacturer Code:MA1159
Catalog # PIMA1159
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 13A9 |
Immunogen | MBP fusion protein containing the full cytoplasmic domain of human N-cadherin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cadherin 2, type 1, N-cadherin (neuronal); Cadherin-2; Cadherin-2 CDw325 CD325 Neural cadherin Calcium-Dependent Adhesion Protein; calcium-dependent adhesion protein, neuronal; CD325; CDw325; N-cadherin; N-cadherin 1; Neural cadherin
Gene Aliases: CD325; CDH2; CDHN; CDw325; N-cadherin; NCAD
UniProt ID: (Human) P19022, (Mouse) P15116
Entrez Gene ID: (Human) 1000, (Mouse) 12558
Molecular Function: cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.