Invitrogen
Manufacturer Code:PA519486
Catalog # PIPA519486
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 800 - 900 of Human N Cadherin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C or -80°C if preferred |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cadherin 2 type 1 N-cadherin (neuronal); cadherin 2, type 1, N-cadherin (neuronal); cadherin 2, type 1, N-cadherin (neuronal) L homeolog; cadherin 2, type 1, N-cadherin a (neuronal); Cadherin-2; Cadherin-2 Neural-cadherin CDw325 antigen CD325 CDHN NCAD Cadherin-N; Cadherin-2A; calcium-dependent adhesion protein, neuronal; CD325; CDw325; N-cadherin; N-cadherin 1; N-cadherin A; N-cadherin-2; Neural cadherin; Neural cadherin A; Neural cadherin-2
Gene Aliases: cadherin-2; CD325; CDH2; cdh2-a; cdh2-b; cdh2a; CDHN; cdhn-a; CDw325; N-Cad; N-cadherin; NCAD; XELAEV_18031953mg
UniProt ID: (Human) P19022, (Mouse) P15116, (Rat) Q9Z1Y3, (Xenopus) P33147
Entrez Gene ID: (Pig) 100515322, (Human) 1000, (Mouse) 12558, (Rat) 83501, (Xenopus) 378518
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.