Invitrogen
Manufacturer Code:PA517526
Catalog # PIPA517526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Two synthetic peptides corresponding to two conserved regions of human N-Cadherin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cadherin 2 type 1 N-cadherin (neuronal); cadherin 2, neuronal; cadherin 2, type 1, N-cadherin (neuronal); Cadherin-2; calcium-dependent adhesion protein, neuronal; CD325; CDw325; cell adhesion molecule; glass onion; labyrinth; N-cadherin; N-cadherin 1; Neural cadherin; Parachute; ZNCAD
Gene Aliases: bib; CD325; CDH2; CDHN; CDw325; glo; lyr; N-cadherin; NCAD; pac; wu:fb47h04
UniProt ID: (Human) P19022, (Mouse) P15116, (Rat) Q9Z1Y3, (Zebrafish) Q90275
Entrez Gene ID: (Human) 1000, (Bovine) 281062, (Mouse) 12558, (Rat) 83501, (Zebrafish) 30291
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.