Novus Biologicals
Manufacturer Code:NBP294531100UL
Catalog # NB023586
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 49-178 of human NAA20 (NP_057184.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dJ1002M8.1 dJ1175I6.2 (N-terminal acetyltransferase complex ard1 subunit) EC 2.3.1.88 homolog) N(alpha)-acetyltransferase 20 NatB catalytic subunit N-acetyltransferase 3 homolog N-acetyltransferase 5 N-acetyltransferase 5 (ARD1 homolog S. cerevisiae) N-acetyltransferase 5 (GCN5-related putative) N-acetyltransferase 5 ARD1 subunit (arrest-defective 1 S. cerevisiae N-alpha-acetyltransferase 20 NatB catalytic subunit NAT3N-terminal acetyltransferase B complex catalytic subunit NAT5 NAT5dJ1002M8.1 (N-terminal acetyltransferase complex ard1 subunit) NatB complex subunit NAT5 N-terminal acetyltransferase B complex catalytic subunit NAA20 N-terminal acetyltransferase complex ARD1 subunit; Methionine N-acetyltransferase; N-acetyltransferase 3 homolog; N-acetyltransferase 5; N-acetyltransferase 5 (ARD1 homolog, S. cerevisiae); N-acetyltransferase 5 (GCN5-related, putative); N-acetyltransferase 5, ARD1 subunit (arrest-defective 1, S. cerevisiae, homolog); N-alpha-acetyltransferase 20; N-alpha-acetyltransferase 20, NatB catalytic subunit; N-terminal acetyltransferase B complex catalytic subunit NAA20; N-terminal acetyltransferase B complex catalytic subunit NAT5; N-terminal acetyltransferase complex ARD1 subunit; NatB catalytic subunit; NatB complex subunit NAT5
Gene Aliases: dJ1002M8.1; NAA20; NAT3; NAT3P; NAT5; NAT5P
UniProt ID: (Human) P61599
Entrez Gene ID: (Human) 51126
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.