Invitrogen
Manufacturer Code:PA528861
Catalog # PIPA528861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 139 of Human N-Ras |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTPase NRas; N-RAS Neuroblastoma RAS NRAS NRAS1 NS6 ALPS4 Neuroblastoma RAS viral (v-ras) oncogene homolog RAS RASN GTPase Nras Transforming protein N-Ras; transforming protein N-Ras
Gene Aliases: AV095280; N-ras
Entrez Gene ID: (Mouse) 18176
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.