Novus Biologicals
Manufacturer Code:NBP148309SS
Catalog # NBP148309SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 13A9 |
Immunogen | Cytoplasmic domain of human N Cadherin [Swiss-Prot# P19022] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cadherin 2 N-cadherin (neuronal) cadherin 2 type 1 N-cadherin (neuronal) cadherin-2 CD325 CD325 antigen CDHNcalcium-dependent adhesion protein neuronal CDw325 N-cadherin NCADN-cadherin 1 Neural cadherin neural-cadherin; cadherin 2, type 1, N-cadherin (neuronal); Cadherin-2; calcium-dependent adhesion protein, neuronal; CD325; CDw325; N-cadherin; N-cadherin 1; Neural cadherin
Gene Aliases: CD325; CDH2; CDHN; CDw325; NCAD
UniProt ID: (Human) P19022
Entrez Gene ID: (Human) 1000
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.