Novus Biologicals
Manufacturer Code:NBP29477020UL
Catalog # NB024057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human GNE (NP_001121699.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bifunctional UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase; bifunctional UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase DMRV GLCNENM glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase IBM2 N-acylmannosamine kinase Uae1 UDP-GlcNAc-2-epimerase/ManAc kinase UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase UDP-N-acetylglucosamine-2-epimerase/N-acetylmannosamine kinase; ManAc kinase; N-acetylmannosamine kinase; N-acylmannosamine kinase; UDP-GlcNAc-2-epimerase; UDP-GlcNAc-2-epimerase/ManAc kinase; UDP-N-acetylglucosamine 2-epimerase (hydrolyzing); UDP-N-acetylglucosamine 2-epimerase/N-acetylmannosamine kinase; UDP-N-acetylglucosamine-2-epimerase/N-acetylmannosamine kinase; Uridine diphosphate-N-acetylglucosamine-2-epimerase
Gene Aliases: DMRV; GLCNE; GNE; IBM2; NM; Uae1
UniProt ID: (Human) Q9Y223
Entrez Gene ID: (Human) 10020
Molecular Function:
carbohydrate kinase
epimerase/racemase
isomerase
kinase
transcription factor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.