Novus Biologicals
Manufacturer Code:NBP100190
Catalog # NBP100190
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-TTHNLEDHTKLP corresponding to internal region according to NP_000503.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Chondroitinase; chondroitinase chondroitinsulfatase EC 3.1.6 EC 3.1.6.4 FLJ17434 FLJ42844 FLJ98217 galactosamine (N-acetyl)-6-sulfate sulfatase Galactose-6-sulfate sulfatase GALNAC6S GalNAc6S sulfatase GAS MPS4A N-acetylgalactosamine-6-sulfatase N-acetylgalactosamine-6-sulfate sulfatase; Chondroitinsulfatase; galactosamine (N-acetyl)-6-sulfate sulfatase; Galactose-6-sulfate sulfatase; GalN6S; GalNAc6S sulfatase; N-acetylgalactosamine-6-sulfatase; N-acetylgalactosamine-6-sulfate sulfatase
Gene Aliases: GalN6S; GALNAC6S; GALNS; GAS; MPS4A
UniProt ID: (Human) P34059
Entrez Gene ID: (Human) 2588
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.