Novus Biologicals
Manufacturer Code:NBP222554AF750
Catalog # NP2224A70
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4D12 |
Immunogen | Recombinant human NAGK (1-344aa) purified from E. coli |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 2.7.1.59 GNKGlcNAc kinase N-acetyl-D-glucosamine kinase N-acetylglucosamine kinaseHSA242910; GlcNAc kinase; N-acetyl-D-glucosamine kinase; N-acetylglucosamine kinase
Gene Aliases: GNK; HSA242910; NAGK
UniProt ID: (Human) Q9UJ70
Entrez Gene ID: (Human) 55577
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.