Invitrogen
Manufacturer Code:PA554070
Catalog # PIPA554070
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Myotrophin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Granule cell differentiation protein; growth factor; MTPN; Myotrophin; Protein V-1; stimulates myocyte growth; V-1 protein
Gene Aliases: 5033418D15Rik; GCDP; MTPN; V-1; V1
UniProt ID: (Human) P58546, (Rat) P62775, (Mouse) P62774
Entrez Gene ID: (Human) 136319, (Rat) 79215, (Mouse) 14489
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.