Novus Biologicals
Manufacturer Code:NBP154959
Catalog # NBP154959
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to MTPN(myotrophin) The peptide sequence was selected from the middle region of MTPN (NP_665807). Peptide sequence GRKPLHYAADCGQLEILEFLLLKGADINAPDKHHITPLLSAVYEGHVSCV. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ31098 FLJ99857 GCDP granule cell differentiation protein myotrophin Protein V-1 V-1; granule cell differentiation protein; Myotrophin; Protein V-1
Gene Aliases: GCDP; MTPN; V-1
UniProt ID: (Human) P58546
Entrez Gene ID: (Human) 136319
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.