Invitrogen
Manufacturer Code:PA511936
Catalog # PIPA511936
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 9-38 amino acids from the N-terminal region of human Myostatin (GDF8) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 2°-8°C for up to 6 months. For long term storage store at -20°C |
Myostatin Antibody, PA5-11936, from Invitrogen™. Species Reactivity: Human, Mouse; Applications: Immunohistochemistry, Western Blot.
For Research Use Only
Protein Aliases: compact; GDF-8; growth differentiation factor 8; Growth/differentiation factor 8; Growth/differentiation factor 8 Myostatin; Myostatin
Gene Aliases: Cmpt; GDF8; MSLHP; MSTN
UniProt ID: (Human) O14793, (Mouse) O08689
Entrez Gene ID: (Human) 2660, (Mouse) 17700
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.