Invitrogen
Manufacturer Code:PA5114224
Catalog # PIPA5114224
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal region of Mouse Mstn |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cmpt compact GDF8 GDF-8 growth differentiation factor 8 Growth/differentiation factor 8 H-Myo-Propeptide H-Myostatin MSLHP Mstn Myostatin; compact; GDF-8; growth differentiation factor 8; Growth/differentiation factor 8; Myostatin
Gene Aliases: Cmpt; Gdf8; Mstn
UniProt ID: (Mouse) O08689
Entrez Gene ID: (Mouse) 17700
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.