Invitrogen
Manufacturer Code:MA511748
Catalog # PIMA511748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MYSN02 (MY-32) |
Immunogen | Rabbit muscle myosin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: epididymis luminal protein 71; fast 2a myosin heavy chain; inclusion body myopathy 3, autosomal dominant; MyHC-2a; MyHC-2x; MyHC-IIa; MyHC-IIx/d; myHC-IIx/d myosin heavy chain 1 myosin heavy chain 2x myosin heavy chain IIx/d myosin heavy chain skeletal muscle adult 1 myosin heavy polypeptide 1 skeletal muscle adult myosin-1; Myosin heavy chain 1; Myosin heavy chain 2; Myosin heavy chain 2a; Myosin heavy chain 2x; Myosin heavy chain IIa; Myosin heavy chain IIx/d; Myosin heavy chain, skeletal muscle, adult 1; Myosin heavy chain, skeletal muscle, adult 2; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy chain 2, skeletal muscle, adult; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin, heavy polypeptide 2, skeletal muscle, adult; Myosin-1; Myosin-2; type IIA myosin heavy chain
Gene Aliases: HEL71; IBM3; MYH1; MYH2; MYH2A; MYHa; MYHas8; MyHC-2A; MyHC-2x; MyHC-2X/D; MyHC-IIa; MYHSA1; MYHSA2; MYPOP
UniProt ID: (Human) P12882, (Human) Q9UKX2
Entrez Gene ID: (Human) 4619, (Human) 4620
Molecular Function: G-protein modulator actin binding motor protein actin family cytoskeletal protein cell junction protein cytoskeletal protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.