Invitrogen
Manufacturer Code:PA531466
Catalog # PIPA531466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 51 and 273 of Human MYH1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MyHC-2x; MYHSA1 MYHa MyHC-2X/D MyHC-2x myHC-IIx/d myosin heavy chain 1 myosin heavy chain 2x myosin heavy chain IIx/d myosin heavy chain skeletal muscle adult 1 myosin heavy polypeptide 1 skeletal muscle adult myosin-1; Myosin heavy chain 1; myosin heavy chain 2A; Myosin heavy chain 2x; Myosin heavy chain, skeletal muscle, adult 1; Myosin-1; myosin-2; skeletal muscle myosin heavy chain II-a
Gene Aliases: A530084A17Rik; IId; IId/x; MdMs; MHC-2X/D; MHC2A; MHC2X/D; Myh1; Myh2a; MyHC-IIa; MyHC-IId/x; MYHC-IIX; MyHC-IIx/d; Myhs-f; Myhs-f1; Myhs-f2; Myhsf1; Myhsf2
UniProt ID: (Mouse) Q5SX40
Entrez Gene ID: (Mouse) 17879, (Mouse) 17882
Molecular Function: G-protein modulator actin binding motor protein actin family cytoskeletal protein cell junction protein cytoskeletal protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.