Invitrogen
Manufacturer Code:PA128037
Catalog # PIPA128037
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Full length native protein (purified) (Human). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2A myosin heavy chain; epididymis luminal protein 71; fast 2a myosin heavy chain; inclusion body myopathy 3, autosomal dominant; MYH1 MYH2 myosin heavy and light chain MyHC-2x Myosin-1 myosin skeletal muscle; MyHC-2a; MyHC-2x; MyHC-IIa; MyHC-IIx/d; Myosin heavy chain 1; Myosin heavy chain 2; Myosin heavy chain 2a; Myosin heavy chain 2x; Myosin heavy chain IIa; Myosin heavy chain IIx/d; myosin heavy chain type IIx; Myosin heavy chain, skeletal muscle, adult 1; Myosin heavy chain, skeletal muscle, adult 2; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy chain 2, skeletal muscle, adult; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin, heavy polypeptide 2, skeletal muscle, adult; Myosin-1; Myosin-2; skeletal muscle myosin heavy chain II-a; type 2X myosin heavy chain; type IIA myosin heavy chain
Gene Aliases: A530084A17Rik; HEL71; IBM3; IId; IId/x; MdMs; MHC-2X/D; MHC2A; MHC2X/D; MYH1; MYH2; MYH2A; MYHa; MYHas8; MYHC; MyHC-2A; MyHC-2x; MyHC-2X/D; MyHC-IIA; MyHC-IId/x; MYHC-IIX; MyHC-IIx/d; Myhs-f; Myhs-f1; Myhs-f2; MYHSA1; MYHSA2; Myhsf1; Myhsf2; MYPOP
UniProt ID: (Human) P12882, (Human) Q9UKX2, (Mouse) Q5SX40
Entrez Gene ID: (Human) 4619, (Human) 4620, (Mouse) 17879, (Rat) 287408, (Rat) 691644, (Mouse) 17882
Molecular Function:
G-protein modulator
actin binding motor protein
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.