Novus Biologicals
Manufacturer Code:NBP2296800.025ML
Catalog # NB019426
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SA19 |
Immunogen | KLH-conjugated synthetic peptide containing the sequence.RS[pT]GV. in which [pT] corresponds to phosphorylated threonine at amino acid residue 850 of chicken myosin phosphatase target subunit 1 (MYPT1). Corresponds to Thr853 in human MYPT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: MBSmyosin phosphatase target subunit 1 MGC133042 Myosin phosphatase target subunit 1 Myosin phosphatase-targeting subunit 1 MYPT1protein phosphatase 1 regulatory subunit 12A protein phosphatase 1 regulatory (inhibitor) subunit 12A Protein phosphatase myosin-binding subunit; myosin binding subunit; Myosin phosphatase target subunit 1; myosin phosphatase, target subunit 1; Myosin phosphatase-targeting subunit 1; Protein phosphatase 1 regulatory subunit 12A; protein phosphatase 1, regulatory (inhibitor) subunit 12A; protein phosphatase 1, regulatory subunit 12A; Protein phosphatase myosin-binding subunit
Gene Aliases: M130; MBS; MYPT1; PPP1R12A
UniProt ID: (Human) O14974
Entrez Gene ID: (Human) 4659
Molecular Function: enzyme modulator phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.