Proteintech
Manufacturer Code:602291IG150UL
Catalog # 50-173-6330
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5A1F1 |
Immunogen | Myosin Light Chain 2 Fusion Protein Ag1356 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cardiac myosin light chain 2; cardiac myosin light chain-2; cardiac ventricular myosin light chain 2; MLC 2 MLC 2v MLC2 MLC2v MYL2 Myosin Light Chain 2; MLC-2; MLC-2s/v; MLC-2V; Myosin light chain 2, slow skeletal/ventricular muscle isoform; myosin light chain 2V; myosin light chain, phosphorylatable, cardiac ventricles; myosin light chain-2; Myosin regulatory light chain 2, ventricular/cardiac muscle isoform; myosin, light chain 2, regulatory, cardiac, slow; myosin, light polypeptide 2, regulatory, cardiac, slow; regulatory light chain of myosin; RLC of myosin; slow cardiac myosin regulatory light chain 2; Ventricular myosin light chain 2; ventricular myosin regulatory light chain
Gene Aliases: CMH10; MLC-2; MLC-2s/v; MLC-2v; MLC2; MLC2V; MYL2; Mylpc
UniProt ID: (Human) P10916, (Rat) P08733, (Mouse) Q9QVP3
Entrez Gene ID: (Human) 4633, (Pig) 396690, (Rat) 363925, (Mouse) 17906
Molecular Function: actin family cytoskeletal protein calcium-binding protein calmodulin cytoskeletal protein intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.