Novus Biologicals
Manufacturer Code:NBP149858
Catalog # NBP149858
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-ETKQLELDLN corresponding to internal region according to NP_001135967.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Dilute myosin heavy chain non-muscle GS1myosin heavy polypeptide kinase MYH12myosin-12 MYO5 Myosin heavy chain 12 myosin V myosin VA (heavy chain 12 myoxin) myosin VA (heavy polypeptide 12 myoxin) Myosin-12 myosin-Va Myoxin MYR12; Dilute myosin heavy chain, non-muscle; Myosin heavy chain 12; myosin V; myosin VA (heavy chain 12, myoxin); myosin, heavy polypeptide kinase; Myosin-12; myosin-Va; Myoxin; Unconventional myosin-Va
Gene Aliases: GS1; MYH12; MYO5; MYO5A; MYR12
UniProt ID: (Human) Q9Y4I1
Entrez Gene ID: (Human) 4644
Molecular Function: G-protein modulator actin binding motor protein actin family cytoskeletal protein cell junction protein cytoskeletal protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.