Invitrogen
Manufacturer Code:14650382
Catalog # 50-112-2708
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MF20 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2B myosin heavy chain; fast myosin heavy chain 2B; MYH2B MyHC-2B MyHC-IIb; myh4; MyHC IIb isoform; MyHC-2b; MyHC-IIb; Myosin heavy chain 2b; Myosin heavy chain 4; Myosin heavy chain IIb; myosin heavy chain type IIb; Myosin heavy chain, skeletal muscle; Myosin heavy chain, skeletal muscle, fetal; myosin, heavy chain 4, skeletal muscle; myosin, heavy polypeptide 4, skeletal muscle; Myosin-4; unnamed protein product
Gene Aliases: AI506973; MHC2B; Minimsc; Minmus; MM; MYH-2B; MYH2B; MYH4; MyHC-2B; MyHC-IIb; Myhsf
UniProt ID: (Human) Q9Y623, (Dog) Q076A5, (Mouse) Q5SX39, (Rat) Q29RW1
Entrez Gene ID: (Human) 4622, (Bovine) 408020, (Dog) 479502, (Rabbit) 100009548, (Guinea pig) 100726456, (Mouse) 17884, (Rat) 360543
Molecular Function:
G-protein modulator
actin binding motor protein
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.