Invitrogen
Manufacturer Code:14650337
Catalog # 14-650-337
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MF20 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2B myosin heavy chain; 2B myosin heavy chain AI506973 fast myosin heavy chain 2B LOC100009548 LOW QUALITY PROTEIN: myosin-4 MHC MHC2B Minimsc Minmus MM MYH2B MYH-2B myh4 MyHC IIb isoform myHC-2b MyHC-IIb Myhsf myosin heavy chain myosin heavy chain 2b Myosin heavy chain 4 myosin heavy chain IIB myosin heavy chain IIb isoform myosin heavy chain isoform 2b myosin heavy chain type IIb Myosin heavy chain skeletal muscle myosin heavy chain skeletal muscle fetal Myosin heavy chain skeletal muscle juvenile myosin heavy chain 4 skeletal muscle myosin heavy polypeptide 4 skeletal muscle myosin-4; fast myosin heavy chain 2B; myh4; MyHC IIb isoform; MyHC-2b; MyHC-IIb; Myosin heavy chain 2b; myosin heavy chain 2X; Myosin heavy chain 4; Myosin heavy chain IIb; myosin heavy chain type IIb; Myosin heavy chain, skeletal muscle; Myosin heavy chain, skeletal muscle, fetal; Myosin heavy chain, skeletal muscle, juvenile; myosin, heavy chain 4, skeletal muscle; myosin, heavy polypeptide 4, skeletal muscle; Myosin-4; unnamed protein product
Gene Aliases: AI506973; MHC; MHC2B; Minimsc; Minmus; MM; MYH-2B; MYH1; MYH2B; MYH4; MyHC-2B; MyHC-IIb; Myhsf
UniProt ID: (Human) Q9Y623, (Dog) Q076A5, (Rabbit) Q28641, (Mouse) Q5SX39, (Rat) Q29RW1
Entrez Gene ID: (Human) 4622, (Bovine) 408020, (Dog) 479502, (Rabbit) 100009548, (Rabbit) 100125991, (Guinea pig) 100726456, (Mouse) 17884, (Rat) 360543
Molecular Function:
G-protein modulator
actin binding motor protein
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.