Invitrogen
Manufacturer Code:PA5116616
Catalog # PIPA5116616
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1450-1550 of human MYH4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2B myosin heavy chain; 2B myosin heavy chain AI506973 fast myosin heavy chain 2B LOC100009548 LOW QUALITY PROTEIN: myosin-4 MHC MHC2B Minimsc Minmus MM MYH2B MYH-2B myh4 MyHC IIb isoform myHC-2b MyHC-IIb Myhsf myosin heavy chain myosin heavy chain 2b Myosin heavy chain 4 myosin heavy chain IIB myosin heavy chain IIb isoform myosin heavy chain isoform 2b myosin heavy chain type IIb Myosin heavy chain skeletal muscle myosin heavy chain skeletal muscle fetal Myosin heavy chain skeletal muscle juvenile myosin heavy chain 4 skeletal muscle myosin heavy polypeptide 4 skeletal muscle myosin-4; myh4; MyHC-2b; MyHC-IIb; Myosin heavy chain 2b; Myosin heavy chain 4; Myosin heavy chain IIb; myosin heavy chain type IIb; Myosin heavy chain, skeletal muscle, fetal; myosin, heavy chain 4, skeletal muscle; myosin, heavy polypeptide 4, skeletal muscle; Myosin-4
Gene Aliases: AI506973; MHC2B; Minimsc; Minmus; MM; MYH-2B; MYH2B; MYH4; MyHC-2B; MyHC-IIb; Myhsf
UniProt ID: (Human) Q9Y623, (Mouse) Q5SX39, (Rat) Q29RW1
Entrez Gene ID: (Human) 4622, (Mouse) 17884, (Rat) 360543
Molecular Function: G-protein modulator actin binding motor protein actin family cytoskeletal protein cell junction protein cytoskeletal protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.