R&D Systems
Manufacturer Code:MAB6686SP
Catalog # MAB6686SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 671038 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Myogenin Ser124-Asn224 Accession # P15173 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bHLHc3; BHLHC3 bHLHc3Myogenic factor-4 myogenin Class C basic helix-loop-helix protein 3 myf-4 MYF4 MYF4myogenin MYOG Myogenic factor 4 MYOGENIN myogenin (myogenic factor 4); Class C basic helix-loop-helix protein 3; Myf-4; Myogenic factor 4; Myogenin
Gene Aliases: BHLHC3; myf-4; MYF4; MYOG
UniProt ID: (Human) P15173
Entrez Gene ID: (Human) 4656
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.