Invitrogen
Manufacturer Code:MA141042
Catalog # 01-676-450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5FD |
Immunogen | Recombit GST tagged fusion protein corresponding to residues 30-224 of rat myogenin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bHLHc3; Class C basic helix-loop-helix protein 3; Myf-4; MYF4 MYOGENIN bHLHc3; MYOD1-related protein; Myogenic factor 4; Myogenin
Gene Aliases: BHLHC3; myf-4; MYF4; myo; MYOG
UniProt ID: (Human) P15173, (Mouse) P12979, (Rat) P20428
Entrez Gene ID: (Human) 4656, (Mouse) 17928, (Rat) 29148
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.