Invitrogen
Manufacturer Code:PA519290
Catalog # PIPA519290
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (TDAHNLHSLTSIVD) corresponding to the C-terminus amino acids of MYOG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: MYOD1-related protein; MYOG myogenin (myogenic factor 4) MYF4 MYOGENIN bHLHc3 Myogenic factor-4 myogenin myogenic factor 4 myogenin; Myogenin
Gene Aliases: bHLHc3; MYF4; myo; Myog
UniProt ID: (Mouse) P12979
Entrez Gene ID: (Mouse) 17928
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.