Novus Biologicals
Manufacturer Code:NBP237656
Catalog # NBP237656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1C8 |
Immunogen | Purified recombinant fragment of human MYOD1 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: bHLHc1; bHLHc1BHLHC1 Class C basic helix-loop-helix protein 1 MYF3Myf-3 MYODmyoblast determination protein 1 myogenic differentiation 1 Myogenic factor 3myf-3 PUM; Class C basic helix-loop-helix protein 1; Myf-3; Myoblast determination protein 1; Myogenic factor 3
Gene Aliases: BHLHC1; MYF3; MYOD; MYOD1; PUM
UniProt ID: (Human) P15172
Entrez Gene ID: (Human) 4654
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.