Novus Biologicals
Manufacturer Code:NBP2443590.1MG
Catalog # NBP24435901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MYADM/972 |
Immunogen | Recombinant human MYADM protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MYADM Myeloid Upregulated Protein Protein SB135 SB135; myeloid upregulated protein; Myeloid-associated differentiation marker; Protein SB135
Gene Aliases: MYADM; SB135; UNQ553/PRO1110
UniProt ID: (Human) Q96S97
Entrez Gene ID: (Human) 91663
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.