Novus Biologicals
Manufacturer Code:NB1001608
Catalog # NB1001608
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chickens were immunized with a synthetic peptide/keyhole limpet hemocyanin (KLH) conjugate. The peptide corresponded to a region of the PLP gene product shared between the human (NP_000524 PLP1 isoform 1 and PLP1 isoform 2 (NP_955772)] and mouse (P60202). After |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: HLD1 lipophilin major myelin proteolipid protein MMPL myelin proteolipid protein PLP PLP/DM20 PMD proteolipid protein 1 spastic paraplegia 2 uncomplicated SPG2; Lipophilin; major myelin proteolipid protein; Myelin proteolipid protein; PLP
Gene Aliases: GPM6C; HLD1; MMPL; PLP; PLP/DM20; PLP1; PMD; SPG2
UniProt ID: (Human) P60201
Entrez Gene ID: (Human) 5354
Molecular Function:
myelin protein
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.