Invitrogen
Manufacturer Code:MA190154
Catalog # PIMA190154
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | IOA3 |
Immunogen | Recombit v-myb protein DNA binding protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: gag-myb protein; myb proto-oncogene protein; myeloblastosis proto-oncogene product; oncogene AMV; Proto-oncogene c-Myb; RP1-32B1.1 Cmyb c-myb c-myb&lowbarCDS efg; Transcriptional activator Myb; tumor-specific myb protein; v-myb avian myeloblastosis viral oncogene homolog
Gene Aliases: AI550390; c-myb; c-myb_CDS; Cmyb; efg; M16449; MYB
UniProt ID: (Human) P10242, (Mouse) P06876
Entrez Gene ID: (Human) 4602, (Mouse) 17863
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.