Invitrogen
Manufacturer Code:PA586288
Catalog # PIPA586288
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the N-terminus of Human c-Myb. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gag-myb protein; myb proto-oncogene protein; myb-related protein B; myeloblastosis proto-oncogene product; oncogene AMV; Proto-oncogene c-Myb; Transcriptional activator Myb; Transcriptional activator Myb Proto-oncogene c-Myb MYB; tumor-specific myb protein; v-myb avian myeloblastosis viral oncogene homolog
Gene Aliases: AI550390; c-myb; c-myb_CDS; Cmyb; efg; M16449; MYB
UniProt ID: (Human) P10242, (Mouse) P06876
Entrez Gene ID: (Human) 4602, (Mouse) 17863, (Rat) 296344
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.