Invitrogen
Manufacturer Code:PA518744
Catalog # PIPA518744
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (QRHYNDEDPEKEKR) corresponding to the internal amino acids of MYB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: c-myb MYB v-myb myeloblastosis viral oncogene homolog (avian) c-myb&lowbarCDS efg Avian myeloblastosis viral (v-myb) oncogene homolog c-myb protein (140 AA) c-myb10A&lowbarCDS c-myb13A&lowbarCDS c-myb14A&lowbarCDS c-myb8B&lowbarCDS v-myb avian myeloblastosis viral oncogene homolog v-myb myeloblastosis viral oncogene homolog; oncogene AMV; Proto-oncogene c-Myb; Transcriptional activator Myb; v-myb avian myeloblastosis viral oncogene homolog
Gene Aliases: c-myb; c-myb_CDS; Cmyb; efg; MYB
UniProt ID: (Human) P10242
Entrez Gene ID: (Human) 4602
Molecular Function:
DNA binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.