R&D Systems
Manufacturer Code:MAB7199SP
Catalog # MAB7199SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 751631 |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse MyBPC3 Pro2-Ser169 Accession # O70468 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: C-protein, cardiac muscle isoform; Cardiac MyBP-C; Cardiac MyBP-C CMH4 C-protein cardiac muscle isoform DKFZp779E1762 FHC MYBP-C myosin binding protein C cardiac myosin-binding protein C cardiac myosin-binding protein C cardiac-type; Myosin-binding protein C, cardiac-type; truncated cardiac myosin-binding protein C
Gene Aliases: CMD1MM; CMH4; FHC; LVNC10; MYBP-C; MYBPC3
UniProt ID: (Human) Q14896
Entrez Gene ID: (Human) 4607
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.