Novus Biologicals
Manufacturer Code:NBP289028
Catalog # NB036619
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to aa 276-288 (G276AGRRTpSDSHEDA288) of MyBPC3 (cardiac type) (cMyBP-C) phosphorylated at Ser282. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-protein, cardiac muscle isoform; Cardiac MyBP-C; Cardiac MyBP-C CMH4 C-protein cardiac muscle isoform DKFZp779E1762 FHC MYBP-C myosin binding protein C cardiac myosin-binding protein C cardiac myosin-binding protein C cardiac-type; Myosin-binding protein C, cardiac-type; truncated cardiac myosin-binding protein C
Gene Aliases: CMD1MM; CMH4; FHC; LVNC10; MYBP-C; MYBPC3
UniProt ID: (Human) Q14896
Entrez Gene ID: (Human) 4607
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.