R&D Systems
Manufacturer Code:MAB7687
Catalog # MAB7687
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 842735 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Muscle Phosphofructokinase/PFKM Asn674-Val780 Accession # P08237 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 6-phosphofructo-1-kinase; 6-phosphofructokinase muscle type EC 2.7.16-phosphofructo-1-kinase EC 2.7.1.11 GSD7 MGC8699 PFK1 PFK-1 PFKA PFK-A PFKM PFKX Phosphofructo-1-kinase isozyme A Phosphofructokinase 1 phosphofructokinase muscle phosphofructokinase polypeptide X Phosphofructokinase-1 Phosphofructokinase-M phosphohexokinase; 6-phosphofructokinase type A; 6-phosphofructokinase, muscle type; ATP-dependent 6-phosphofructokinase, muscle type; ATP-PFK; PFK-A; Phosphofructo-1-kinase isozyme A; phosphofructokinase 1; phosphofructokinase muscle; phosphofructokinase, polypeptide X; phosphofructokinase-M; Phosphohexokinase; protein phosphatase 1, regulatory subunit 122
Gene Aliases: ATP-PFK; GSD7; PFK-1; PFK1; PFKA; PFKM; PFKX; PPP1R122
UniProt ID: (Human) P08237
Entrez Gene ID: (Human) 5213
Molecular Function: carbohydrate kinase kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.