Novus Biologicals
Manufacturer Code:NBP232169R
Catalog # NBP232169R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 1 to 50 of human Phosphofructokinase Muscle using the numbering given in entry NP_000280.1 (GeneID 5213). |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6-phosphofructo-1-kinase; 6-phosphofructokinase muscle type EC 2.7.16-phosphofructo-1-kinase EC 2.7.1.11 GSD7 MGC8699 PFK1 PFK-1 PFKA PFK-A PFKX Phosphofructo-1-kinase isozyme A Phosphofructokinase 1 phosphofructokinase muscle phosphofructokinase polypeptide X phosphofructokinase-M phosphohexokinase; 6-phosphofructokinase type A; 6-phosphofructokinase, muscle type; ATP-dependent 6-phosphofructokinase, muscle type; ATP-PFK; PFK-A; Phosphofructo-1-kinase isozyme A; phosphofructokinase 1; phosphofructokinase muscle; phosphofructokinase, polypeptide X; phosphofructokinase-M; Phosphohexokinase; protein phosphatase 1, regulatory subunit 122
Gene Aliases: ATP-PFK; GSD7; PFK-1; PFK1; PFKA; PFKM; PFKX; PPP1R122
UniProt ID: (Human) P08237
Entrez Gene ID: (Human) 5213
Molecular Function:
carbohydrate kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.