R&D Systems
Manufacturer Code:FAB10323S100UG
Catalog # FAB10323S10
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 590123 |
Immunogen | NS0 mouse myeloma cell line transfected with human CHRM5 Met1-Pro532 Accession &num P08912 |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor, muscarinic 5; cholinergic receptor muscarinic 5 HM5 M5 M5R mAChR M5 MGC41838 muscarinic acetylcholine receptor M5; cholinergic receptor, muscarinic 5; Muscarinic acetylcholine receptor M5
Gene Aliases: CHRM5; HM5
UniProt ID: (Human) P08912
Entrez Gene ID: (Human) 1133
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.