Novus Biologicals
Manufacturer Code:NB1002064R
Catalog # NB1002064R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | mta30 2g10/3 |
Immunogen | Human MUS81 |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Crossover junction endonuclease MUS81; crossover junction endonuclease MUS81 EC 3.1.22 EC 3.1.22.- FLJ21012 FLJ44872 MUS81 endonuclease homolog (S. cerevisiae) MUS81 endonuclease homolog (yeast) SLX3 SLX3 structure-specific endonuclease subunit homolog; MUS81 endonuclease homolog; SLX3 structure-specific endonuclease subunit homolog
Gene Aliases: MUS81; SLX3
UniProt ID: (Human) Q96NY9
Entrez Gene ID: (Human) 80198
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.