Invitrogen
Manufacturer Code:MA15837
Catalog # 01-674-940
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2G10/3 |
Immunogen | Human Mus81 from Sp2/0 Ag.14 cells |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Crossover junction endonuclease MUS81; FLJ21012 FLJ44872 MUS81; MUS81 endonuclease homolog; SLX3 structure-specific endonuclease subunit homolog
Gene Aliases: MUS81; SLX3
UniProt ID: (Human) Q96NY9
Entrez Gene ID: (Human) 80198
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.