Invitrogen
Manufacturer Code:PA530883
Catalog # PIPA530883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 8 and 222 of Human Munc 13-4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FHL3 HLH3 HPLH3 Munc13-4 protein unc-13 homolog D; likely ortholog of H. sapiens unc-13 homolog D (UNC13D); Munc13-4; Protein unc-13 homolog D
Gene Aliases: 2610108D09Rik; FHL3; HLH3; HPLH3; Jinx; mFLJ00067; Munc13-4; UNC13D
UniProt ID: (Human) Q70J99, (Mouse) B2RUP2
Entrez Gene ID: (Human) 201294, (Mouse) 70450
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.