Novus Biologicals
Manufacturer Code:NB11082375AF647
Catalog # NB11082375Y
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to a C-terminal portion of mouse Mucolipin 1 (within residues 500-580). [Swiss-Prot# Q99J21] |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MG-2; MG-2 ML4TRP-ML1 MLIV MST080 MSTP080 mucolipidin mucolipidosis type IV protein mucolipin 1 mucolipin-1 TRPML1 TRP-ML1 TRPM-L1; ML1; Mucolipidin; mucolipidosis type IV protein; Mucolipin-1; Transient receptor potential channel mucolipin 1; TRPML1
Gene Aliases: MCOLN1; MG-2; ML4; MLIV; MST080; MSTP080; TRP-ML1; TRPM-L1; TRPML1
UniProt ID: (Human) Q9GZU1
Entrez Gene ID: (Human) 57192
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.